missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX11 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92510-0.1ml
This item is not returnable.
View return policy
Description
DDX11 Polyclonal antibody specifically detects DDX11 in Human samples. It is validated for Western Blot
Specifications
| DDX11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CHL1CHL1-related helicase gene-1, CHL1-related protein 1, CHLR1MGC9335, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (CHL1-like helicase homolog, S.cerevisiae), DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 11 (S.cerevisiae CHL1-likehelicase), DEAD/H box protein 11, EC 3.6.1, EC 3.6.1.23, EC 3.6.1.7, EC 3.6.4.13, hCHLR1, Keratinocyte growth factor-regulated gene 2 protein, KRG-2, KRG2CHL1-like helicase homolog, MGC133249, probable ATP-dependent RNA helicase DDX11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human DDX11 (NP_689651.1). MANETQKVGAIHFPFPFTPYSIQEDFMAELYRVLEAGKIGIFESPTGTGKSLSLICGALSWLRDFEQKKREEEARLLETGTGPLHDEKDESLCLSSSCEGAAGTPRPAGEPAWVTQFVQKKEERDLVDRLKAEQARRKQREERLQQLQHRVQLKYAAKRLRQEEEERENLLRLSREMLETGPEAERLEQLESGEEELVLAEYESDEEKKVASRVDEDEDD | |
| 0.1 mL | |
| Cell Biology, Cell Cycle and Replication | |
| 1663 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction