missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX10 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-1576AF488
This item is not returnable.
View return policy
Description
DDX10 Polyclonal antibody specifically detects DDX10 in Human samples. It is validated for Western Blot
Specifications
| DDX10 | |
| Polyclonal | |
| Western Blot | |
| DDX10-NUP98 fusion protein type 2, DEAD (Asp-Glu-Ala-Asp) box polypeptide 10, DEAD box protein 10, DEAD box-10, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 10 (RNA helicase), EC 3.6.1, EC 3.6.4.13, HRH-J8, probable ATP-dependent RNA helicase DDX10 | |
| The immunogen recognized by this antibody maps to a region between residues 500 and 550 of human DEAD (Asp-Glu-Ala-Asp) Box Polypeptide 10 using the numbering given in entry NP_004389.2 (GeneID 1662). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 1662 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction