missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11357-1-AP-150UL
This item is not returnable.
View return policy
Description
DDX1 (ATP-dependent RNA helicase DDX1) acts as an ATP-dependent RNA helicase. It is able to unwind both RNA-RNA and RNA-DNA duplexes. DDX1 possesses 5′ single-stranded RNA overhand nuclease activity and ATPase activity on various RNA, but not DNA polynucleotides. DDX1, together with RELA, acts as a coactivator to enhance NF-kappa-b-mediated transcriptional activation. It also acts as a positive transcriptional regulator of cyclin CCND2 expression. It binds to the cyclin CCND2 promoter region. DDX1 is a component of a multi-helicase-TICAM1 complex that acts as a cytoplasmic sensor of viral double stranded RNA and plays a role in activation of a cascade of antiviral responses including the induction of proinflammatory cytokines via the adapter molecule TICAM1.Specifications
| DDX1 | |
| Polyclonal | |
| Unconjugated | |
| DDX1 | |
| DBP RB, DDX1, DEAD box protein 1, UKVH5d | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 104721, 1653, 84474 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q641Y8, Q91VR5, Q92499 | |
| DDX1 | |
| DDX1 Fusion Protein Ag1887 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction