missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583227
This item is not returnable.
View return policy
Description
Immunogen sequence: LGKSHIRTDD VHAKDNTRPG ANSPEMWSEA IKILKGEYAV RAIKEHKMDQ AIIFCRTKID CDNLEQYFIQ QGGGPDKKGH QFSCVCLHGD Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.
DDX1 (ATP-dependent RNA helicase DDX1) acts as an ATP-dependent RNA helicase. It is able to unwind both RNA-RNA and RNA-DNA duplexes. DDX1 possesses 5' single-stranded RNA overhand nuclease activity and ATPase activity on various RNA, but not DNA polynucleotides. DDX1, together with RELA, acts as a coactivator to enhance NF-kappa-b-mediated transcriptional activation. It also acts as a positive transcriptional regulator of cyclin CCND2 expression. It binds to the cyclin CCND2 promoter region. DDX1 is a component of a multi-helicase-TICAM1 complex that acts as a cytoplasmic sensor of viral double stranded RNA and plays a role in activation of a cascade of antiviral responses including the induction of proinflammatory cytokines via the adapter molecule TICAM1.
Specifications
| DDX1 | |
| Polyclonal | |
| Unconjugated | |
| DDX1 | |
| AA409185; ATP-dependent RNA helicase DDX1; DBP-RB; Ddx1; DEAD (Asp-Glu-Ala-Asp) box helicase 1; DEAD (Asp-Glu-Ala-Asp) box polypeptide 1; DEAD box polypeptide 1; DEAD box protein 1; DEAD box protein retinoblastoma; DEAD box-1; DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1; DEAD/H-box helicase 1; UKVH5d | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 104721, 1653, 84474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q641Y8, Q91VR5, Q92499 | |
| DDX1 | |
| Recombinant protein corresponding to Human DDX1. Recombinant protein control fragment (Product #RP-95988). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction