missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57379
This item is not returnable.
View return policy
Description
DDX1 Polyclonal specifically detects DDX1 in Human samples. It is validated for Western Blot.
Specifications
| DDX1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ATP-dependent RNA helicase DDX1, DBP-RBDEAD box polypeptide 1, DEAD (Asp-Glu-Ala-Asp) box polypeptide 1, DEAD box protein 1, DEAD box protein retinoblastoma, DEAD box-1, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 1, EC 3.6.1, EC 3.6.1.10, EC 3.6.4.13, UKVH5d | |
| Rabbit | |
| 81 kDa | |
| 100 μL | |
| Cancer, Neuroscience, Transcription Factors and Regulators, Vision | |
| 1653 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q92499 | |
| DDX1 | |
| Synthetic peptides corresponding to DDX1 (DEAD (Asp-Glu-Ala-Asp) box polypeptide 1) The peptide sequence was selected from the C terminal of DDX1. Peptide sequence SQVEPDIKVPVDEFDGKVTYGQKRAAGGGSYKGHVDILAPTVQELAALEK. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 78%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction