missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12389-1-AP-150UL
This item is not returnable.
View return policy
Description
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).Specifications
| DDT | |
| Polyclonal | |
| Unconjugated | |
| DDT | |
| D dopachrome decarboxylase, D dopachrome tautomerase, DDCT, DDT, Phenylpyruvate tautomerase II | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13202, 1652, 29318 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O35215, P30046, P80254 | |
| Ddt | |
| DDT Fusion Protein Ag3065 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction