missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597160
This item is not returnable.
View return policy
Description
Immunogen sequence: MPFLELDTNL PANRVPAGLE KRLCAAAASI LGKPADRVNV TVRPGLAMAL SGSTEPCAQL SISSIGVVGT AEDNRSHSAH FFEFLTKELA LGQDRILIRF FPLESWQIGK IGTVMTFL; Positive Samples: 293T, A-549, Mouse liver, Rat liver; Cellular Location: Cytoplasm.
Tautomerization of D-dopachrome with decarboxylation to give 5,6-dihydroxyindole (DHI).
Specifications
| DDT | |
| Polyclonal | |
| Unconjugated | |
| DDT | |
| C78655; DDCT; D-dopachrome decarboxylase; D-dopachrome tautomerase; Ddt; dopachrome isomerase; hCG_41098; phenylpyruvate tautomerase II | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 13202, 1652, 29318 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 3.36 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O35215, P30046, P80254 | |
| DDT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human DDT (NP_0013461). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction