missing translation for 'onlineSavingsMsg'
Learn More

DDT Antibody [PE], Novus Biologicals™

Product Code. 30737081 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737081 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737081 Supplier Novus Biologicals Supplier No. AF8354P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

DDT Polyclonal antibody specifically detects DDT in Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDT
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias D-dopachrome decarboxylase, D-dopachrome tautomeraseDDCT, EC 4.1.1.84, Phenylpyruvate tautomerase II
Host Species Sheep
Immunogen E. coli-derived recombinant mouse DDT, Pro2-Leu118, Accession # O35215
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1652
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.