missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDRGK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99194-50ul
This item is not returnable.
View return policy
Description
DDRGK1 Polyclonal antibody specifically detects DDRGK1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DDRGK1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| C20orf116, chromosome 20 open reading frame 116, DDRGK domain containing 1, DDRGK domain-containing protein 1, dJ1187M17.3, MGC2592 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DDRGK1. | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 65992 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction