missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDR2 Recombinant Rabbit Monoclonal Antibody (HL1107)

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
DDR2 is a member of a subclass of RTKs and contains a distinct extracellular region encompassing a factor VIII-like domain. DDR1 and DDR2 have been shown to be potently inhibited by Gleevec. DDR2 required for normal bone development. Regulates osteoblast differentiation and chondrocyte maturation via a signaling pathway that involves MAP kinases and leads to the activation of the transcription factor RUNX2. Regulates remodeling of the extracellular matrix by up-regulation of the collagenases MMP1, MMP2 and MMP13, and thereby facilitates cell migration and tumor cell invasion. Promotes fibroblast migration and proliferation, and thereby contributes to cutaneous wound healing.
Specifications
Specifications
| Antigen | DDR2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | HL1107 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Ddr2 |
| Gene Accession No. | Q16832, Q62371 |
| Gene Alias | AW495251; CD_antigen: CD167b; CD167 antigen-like family member B; CD167b; CD167b antigen; cell migration-inducing protein 20; Ddr2; Discoidin domain receptor 2; discoidin domain receptor family, member 2; discoidin domain receptor tyrosine kinase 2; discoidin domain-containing receptor 2; Discoidin domain-containing receptor tyrosine kinase 2; hydroxyaryl-protein; hydroxyaryl-protein kinase; MIG20a; migration-inducing gene 16 protein; neurotrophic tyrosine kinase receptor related 3; neurotrophic tyrosine kinase, receptor-related 3; Ntrkr3; receptor protein-tyrosine kinase TKT; RP11-572K18.1; TKT; TYRO10; tyrosine-protein kinase TYRO 10; tyrosine-protein kinase TYRO10; tyrosylprotein kinase |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?