missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR2 Rabbit anti-Mouse, Clone: 102, Novus Biologicals™
Rabbit Monoclonal Antibody
£296.00 - £449.00
Specifications
| Antigen | DDR2 |
|---|---|
| Clone | 102 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204148
|
Novus Biologicals
NBP2-90632-50UL |
50 μg |
£296.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18032954
|
Novus Biologicals
NBP2-90632-100UL |
100 μg |
£449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDR2 Monoclonal specifically detects DDR2 in Mouse samples. It is validated for ELISA.Specifications
| DDR2 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 4921 | |
| IgG | |
| Protein A purified |
| 102 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
| Recombinant Mouse DDR2 Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title