missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR2 Antibody (290804) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB25381JF525
This item is not returnable.
View return policy
Description
DDR2 Monoclonal antibody specifically detects DDR2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,CyTOF-reported
Specifications
| DDR2 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| CD167 antigen-like family member B, CD167b antigen, Discoidin domain receptor 2, discoidin domain receptor family, member 2, discoidin domain receptor tyrosine kinase 2, discoidin domain-containing receptor 2, EC 2.7.10, EC 2.7.10.1, hydroxyaryl-protein kinase, migration-inducing gene 16 protein, neurotrophic tyrosine kinase receptor related 3, Neurotrophic tyrosine kinase, receptor-related 3, NTRKR3cell migration-inducing protein 20, Receptor protein-tyrosine kinase TKT, TKTMIG20a, TYRO10, Tyrosine-protein kinase TYRO10, tyrosylprotein kinase | |
| Mouse myeloma cell line NS0-derived recombinant human DDR2, Gln24-Arg399, Accession # Q16832 | |
| 0.1 mL | |
| Cardiovascular Biology, CD Markers, Immunology, Neuroscience, Signal Transduction | |
| 4921 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 290804 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido