missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ Human DDR1, GST Tag Recombinant Protein
Klik for at se tilgængelige muligheder
Quantity:
1 mg
100 μg
Pakningsstørrelse:
100µg
Each
Beskrivelse
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
DDR1 is a receptor tyrosine kinase activated by collagen and is reported to regulate adhesion, cell migration and other cellular processes. Receptor tyrosine kinases (RTKs) play a key role in the communication of cells with their microenvironment. These molecules are involved in the regulation of cell growth, differentiation and metabolism. The protein encoded by this gene is a RTK that is widely expressed in normal and transformed epithelial cells and is activated by various types of collagen. This protein belongs to a subfamily of tyrosine kinase receptors with a homology region to the Dictyostelium discoideum protein discoidin I in their extracellular domain. Its autophosphorylation is achieved by all collagens so far tested (type I to type VI). In situ studies and Northern-blot analysis showed that expression of this encoded protein is restricted to epithelial cells, particularly in the kidney, lung, gastrointestinal tract, and brain. In addition, this protein is significantly over-expressed in several human tumors from breast, ovarian, esophageal, and pediatric brain. This gene is located on chromosome 6p21.3 in proximity to several HLA class I genes. Alternative splicing of this gene results in multiple transcript variants.
Tekniske data
Tekniske data
| Accession Number | Q08345 |
| Concentration | See Label |
| For Use With (Application) | Kinase Assay |
| Formulation | proprietary buffer with no preservative |
| Gene ID (Entrez) | 780 |
| Molecular Weight (g/mol) | 78.1 kDa |
| Name | Human DDR1, GST Tag |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Gene Alias | 6030432F18; AI323681; Cak; CD167; CD167 antigen-like family member A; CD167a; Cell adhesion kinase; DAAP-278B20.1; DDR; DDR1; DDR1d; discoidin domain receptor family, member 1; discoidin domain receptor tyrosine kinase 1; discoidin receptor tyrosine kinase; Drd1; Eddr1; Epithelial discoidin domain receptor 1; epithelial discoidin domain-containing receptor 1; HGK2; mammary carcinoma kinase 10; MCK10; MCK-10; Mpk6; NEP; neuroepithelial tyrosine kinase; neurotrophic tyrosine kinase receptor type 4; neurotrophic tyrosine kinase, receptor, type 4; NTRK4; protein-tyrosine kinase 3; Protein-tyrosine kinase 3 A; Protein-tyrosine kinase MPK-6; protein-tyrosine kinase PTK-3; protein-tyrosine kinase RTK-6; PTK3; PTK3A; PTK3A protein tyrosine kinase 3 A; PTK3D; RTK6; TRK E; TRKE; Tyrosine kinase DDR; tyrosine-protein kinase CAK |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion