missing translation for 'onlineSavingsMsg'
Learn More

DDR1/DDR2 [p Tyr796, p Tyr740, p Tyr740] Antibody (1119D) [FITC], Novus Biologicals™

Product Code. 30737862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737862 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737862 Supplier Novus Biologicals Supplier No. FAB25382F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DDR1/DDR2 Monoclonal antibody specifically detects DDR1/DDR2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDR1/DDR2
Applications Western Blot
Classification Monoclonal
Clone 1119D
Conjugate FITC
Formulation PBS
Gene Alias 6030432F18;AI323681;CAK;CD167;CD167a;DDR;DDR1;Drd1;EDDR1;HGK2;MCK10;Mpk6;NEP;NTRK4;PTK3;PTK3A;PTK3D;RTK6;TRKE
Host Species Rabbit
Immunogen Phosphopeptide containing the human DDR2 Y740 site, Accession # Q16832
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.