missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Antibody (Imperial College anti-DDR1) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28204JF585
This item is not returnable.
View return policy
Description
DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DDR1 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
| DDR1/ CD167a | |
| 0.1 mL | |
| Cancer, Cellular Signaling | |
| 780 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Imperial College anti-DDR1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction