missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DDR1 Antibody (Imperial College anti-DDR1) - Humanized, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28204-100ug
This item is not returnable.
View return policy
Description
DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DDR1 | |
| Monoclonal | |
| Unconjugated | |
| Q08345 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Imperial College anti-DDR1 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
| DDR1/ CD167a | |
| 100 μg | |
| Cancer, Cellular Signaling | |
| 780 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction