missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDR1 Antibody (3E3), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81073CL1
This item is not returnable.
View return policy
Description
DDR1 Monoclonal antibody specifically detects DDR1 in Human samples. It is validated for ELISA,Western Blot,Neutralization
Specifications
| DDR1 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD167 antigen-like family member A, CD167a antigen, DDR, discoidin domain receptor family, member 1, discoidin domain receptor tyrosine kinase 1, Discoidin receptor tyrosine kinase, EC 2.7.10, EC 2.7.10.1, EDDR1, ENTRK4, Epithelial discoidin domain receptor 1, HGK2, MCK10, MCK-10, NTRK4, Protein-tyrosine kinase 3A, PTK3A, PTK3A protein tyrosine kinase 3A, receptor, type 4, TRK E | |
| Recombinant human DDR1 extracellular domain. | |
| 0.1 mL | |
| Cancer, Cellular Signaling | |
| 780 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| ELISA, Western Blot, Neutralization | |
| 3E3 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction