missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDP-38003 trihydrochloride, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
DDP-38003 trihydrochloride is an novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM.
Specifications
Specifications
| Chemical Name or Material | DDP-38003 trihydrochloride |
| Physical Form | Solid |
| Color | Reddish Brown |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29Cl3N4O |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 100 mg/mL (217.47 mM; Need ultrasonic) ∣H2O : 50 mg/mL (108.73 mM; Need ultrasonic) |
| SMILES | [H]Cl.[H]Cl.O=C(NC1=CC=C([C@@H]2C[C@H]2N)C=C1)C(C=C3)=CC=C3N4CCN(C)CC4.[H]Cl |
| Molecular Weight (g/mol) | 459.84 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction