missing translation for 'onlineSavingsMsg'
Learn More

DDP-38003 trihydrochloride, MedChemExpress

Product Code. 16477646
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16477646

Brand: Medchem Express HY19612B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DDP-38003 trihydrochloride is an novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM.

DDP-38003 trihydrochloride is an novel, orally available inhibitor of histone lysine-specific demethylase 1A (KDM1A/LSD1) with an IC50 of 84 nM.

Specifications

Chemical Name or Material DDP-38003 trihydrochloride
Physical Form Solid
Color Reddish Brown
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H29Cl3N4O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 100 mg/mL (217.47 mM; Need ultrasonic) ∣H2O : 50 mg/mL (108.73 mM; Need ultrasonic)
SMILES [H]Cl.[H]Cl.O=C(NC1=CC=C([C@@H]2C[C@H]2N)C=C1)C(C=C3)=CC=C3N4CCN(C)CC4.[H]Cl
Molecular Weight (g/mol) 459.84
Formula Weight 459.84
Percent Purity 96.77%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.