missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDOST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561347
This item is not returnable.
View return policy
Description
Immunogen sequence: NYELAVALSR WVFKEEGVLR VGPVSHHRVG ETAPPNAYTV TDLVEYSIVI QQLSNGKWVP FDGDDIQLEF VRIDPFVRTF LKKKGGKYSV QFKLPDVYGV FQFKVDYNRL GYTHLYSSTQ VSVRPLQHTQ Y Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
This gene encodes a component of the oligosaccharyltransferase complex which catalyzes the transfer of high-mannose oligosaccharides to asparagine residues on nascent polypeptides in the lumen of the rough endoplasmic reticulum. The protein complex co-purifies with ribosomes. The product of this gene is also implicated in the processing of advanced glycation end products (AGEs), which form from non-enzymatic reactions between sugars and proteins or lipids and are associated with aging and hyperglycemia.
Specifications
| DDOST | |
| Polyclonal | |
| Unconjugated | |
| DDOST | |
| advanced glycation endproduct receptor 1; advanced glycation end-product receptor 1; AGER1; AGE-R1; CDG1R; Ddost; DDOST 48 kDa subunit; dolichyl-diphosphooligosaccharide protein glycosyltransferase; dolichyl-diphosphooligosaccharide-protein glycosyltransferase; dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase non-catalytic subunit; dolichyl-diphosphooligosaccharide--protein glycosyltransferase subunit (non-catalytic); dolichyl-di-phosphooligosaccharide-protein glycotransferase; dolichyl-diphosphooligosaccharide-protein glycotransferase; KIAA0115; MGC2191; OK/SW-cl.45; OKSWcl45; oligosaccharyl transferase 48 kDa subunit; oligosaccharyltransferase 48 kDa subunit; oligosaccharyltransferase OST48; oligosaccharyltransferase subunit 48; OST; OST48; WBP1; wu:fa11d01; zgc:66068; zgc:66068 protein; zgc:77478 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P39656 | |
| DDOST | |
| Recombinant protein corresponding to Human DDOST. Recombinant protein control fragment (Product #RP-103000). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction