missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDOST Antibody (02), Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £255.00
Specifications
| Antigen | DDOST |
|---|---|
| Clone | 02 |
| Dilution | ELISA 1:1000-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18649958
|
Novus Biologicals
NBP3-05844-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645038
|
Novus Biologicals
NBP3-05844-50ul |
50 μg |
£173.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDOST Monoclonal antibody specifically detects DDOST in Human samples. It is validated for ELISASpecifications
| DDOST | |
| ELISA 1:1000-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 1650 | |
| IgG2a | |
| Protein A purified |
| 02 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| advanced glycation endproduct receptor 1, DDOST 48 kDa subunit, dolichyl-diphosphooligosaccharide-protein glycosyltransferase, dolichyl-diphosphooligosaccharide--protein glycosyltransferase, dolichyl-diphosphooligosaccharide--protein glycosyltransferase 48 kDa subunit, dolichyl-diphosphooligosaccharide-protein glycotransferase, EC 2.4.1.119, KIAA0115MGC2191, Oligosaccharyl transferase 48 kDa subunit, oligosaccharyltransferase 48 kDa subunit, OST48AGER1, OSTOKSWcl45, WBP1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human DDOST (Uniprot#: P39656-1; Ser43-Pro427). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title