missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDO Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557988
This item is not returnable.
View return policy
Description
Immunogen sequence: MLIPHTYPDT PIHTQKQWFR ETFNHLFAIA NSAEAGDAGV HLVSGWQIFQ STPTEEVPFW ADVVLGFRKM TE Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.
DDO is a peroxisomal flavoprotein that catalyzes the oxidative deamination of D-aspartate and N-methyl D-aspartate. Flavin adenine dinucleotide or 6-hydroxyflavin adenine dinucleotide can serve as the cofactor in this reaction.
Specifications
| DDO | |
| Polyclonal | |
| Unconjugated | |
| DDO | |
| 5330420D20Rik; 5730402C02Rik; AI467244; aspartic oxidase; DASOX; D-aspartate oxidase; D-aspartate oxidase, DDO; Ddo; DDO protein; DDO-1; DDO-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8528 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99489 | |
| DDO | |
| Recombinant protein corresponding to Human DDO. Recombinant protein control fragment (Product #RP-95047). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction