missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDIT4L Polyclonal Antibody

Description
The antibody detects endogenous levels of total DDIT4L protein.
DDIT4L inhibits cell growth by regulating the TOR signaling pathway upstream of the TSC1-TSC2 complex and downstream of AKT1.
Specifications
Specifications
| Antigen | DDIT4L |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | DDIT4L |
| Gene Accession No. | Q96D03 |
| Gene Alias | 1700037B15Rik; 1700108M02Rik; Ddit4l; DNA damage inducible transcript 4 like; DNA damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like; DNA-damage-inducible transcript 4-like protein; DNA-damage-inducible transcript 4-like protein-like; HIF-1 responsive protein RTP801-like; homolog of mouse SMHS1; protein regulated in development and DNA damage response 2; REDD2; REDD-2; regulated in development and DNA damage response 2; RTP801L; skeletal muscle hindlimb suspension 1; Smhs1; Soleus muscle atrophied after hindlimb suspension protein 1 |
| Gene Symbols | DDIT4L |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?