missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDIT4L Antibody (DDIT-03), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62218AF532
This item is not returnable.
View return policy
Description
DDIT4L Monoclonal specifically detects DDIT4L in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| DDIT4L | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DDIT4L | |
| N-terminal recombinant fragment of human DDIT4L (amino acids 2-98) | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-18 reacts with CD14, a 53-55 kDa GPI (glycosylphosphatidylinositol)-linked membrane glycoprotein expressed on monocytes, macrophages and weakly on granulocytes; also expressed by most tissue macrophages. In human, the epitope recognized by MEM-18 is located between amino acids 57-64. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| DDIT-03 | |
| Western Blot, Flow Cytometry | |
| DNA damage-inducible transcript 4-like protein, DNA-damage-inducible transcript 4-like, HIF-1 responsive protein RTP801-like, homolog of mouse SMHS1, Protein regulated in development and DNA damage response 2, REDD-2, REDD2Rtp801L, regulated in development and DNA damage response 2, RTP801L | |
| Mouse | |
| Protein A purified | |
| Cancer, mTOR Pathway | |
| 115265 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction