missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DDIT4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583229

Product Code. 16380185

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QLLQESLAQA RLGSRRPARL LMPSQLVSQV GKELLRLAYS EPCGLRGALL DVCVEQGKSC HSVGQLALD Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

DDIT4 gene ontology annotations related to this gene include cellular response to dexamethasone stimulus; intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator; negative regulation of glycolytic process; negative regulation of TOR signaling; protein-containing complex disassembly; reactive oxygen species metabolic process; response to hypoxia.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DDIT4
Polyclonal
Unconjugated
DDIT4
5830413E08Rik; AA415483; DDIT4; Dexamethasone-induced gene 2 protein; dig2; DNA damage inducible transcript 4; DNA damage-inducible transcript 4 protein; DNA-damage-inducible transcript 4; DNA-damage-inducible transcript 4 protein; HIF-1 responsive protein RTP801; HIF-1 responsive RTP801; protein regulated in development and DNA damage response 1; REDD1; REDD-1; RP11-442H21.1; RTP801
Rabbit
Antigen affinity chromatography
RUO
54541
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9NX09
DDIT4
Recombinant protein corresponding to Human DDIT4. Recombinant protein control fragment (Product #RP-95683).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.