missing translation for 'onlineSavingsMsg'
Learn More

DDIT4 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18406682 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406682 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406682 Supplier Novus Biologicals Supplier No. NBP177321AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDIT4 Polyclonal antibody specifically detects DDIT4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDIT4
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formulation 50mM Sodium Borate
Gene Alias Dig2, DNA-damage-inducible transcript 4, FLJ20500, HIF-1 responsive RTP801 (RTP801), Protein regulated in development and DNA damage response 1, REDD-1DNA damage-inducible transcript 4 protein, REDD1HIF-1 responsive protein RTP801, RTP801RP11-442H21.1
Host Species Rabbit
Immunogen Antibody was raised against a 12 amino acid synthetic peptide from near the center of human RTP801. The immunogen is located within amino acids 90 - 140 of RTP801. Amino Acid Squence: QARLGSRRPARL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 54541
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.