missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DDI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114071
This item is not returnable.
View return policy
Description
Immunogen sequence: KNVLVIGTTG TQTYFLPEGE LPLCSRMVSG QDESSDKEIT HSVMDSGRKE For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%.
Ddi1 (DNA-damage inducible 1)appears to release substrate from an ubiquitination complex, making the substrate available for deubiquitination to facilitate proteasomal degradation. Ddi1 could be a potential selective target for lung cancer treatment. Ddi1 runs as a doublet of 54/56 kDa on acrylamide gels besides the calculated MW 44 kDa.
Specifications
| DDI1 | |
| Polyclonal | |
| Unconjugated | |
| DDI1 | |
| 1700011N24Rik; DDI1; DDI1, DNA-damage inducible 1, homolog 1; DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae); DNA damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1; DNA-damage inducible 1 homolog 1 (S. cerevisiae); DNA-damage inducible protein 1; protein DDI1 homolog 1; RGD1559430 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 414301 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WTU0 | |
| DDI1 | |
| Synthetic peptide directed towards the middle region of human DDI1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction