missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDI1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | DDI1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DDI1 Polyclonal antibody specifically detects DDI1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| DDI1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS with 50% glycerol, pH7.3. | |
| 414301 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DDI1, DNA-damage inducible 1, homolog 1, DDI1, DNA-damage inducible 1, homolog 1 (S. cerevisiae), DNA-damage inducible 1 homolog 1 (S. cerevisiae), DNA-damage inducible protein 1, FLJ36017, protein DDI1 homolog 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 97-396 of human DDI1 (NP_001001711.1). SGIAVPGTSSSRPQHPGQQQQRTPAAQRSQGLASGEKVAGLQGLGSPALIRSMLLSNPHDLSLLKERNPPLAEALLSGSLETFSQVLMEQQREKALREQERLRLYTADPLDREAQAKIEEEIRQQNIEENMNIAIEEAPESFGQVTMLYINCKVNGHPLKAFVDSGAQMTIMSQACAERCNIMRLVDRRWAGVAKGVGTQRIIGRVHLAQIQIEGDFLQCSFSILEDQPMDMLLGLDMLRRHQCSIDLKKNVLVIGTTGTQTYFLPEGELPLCSRMVSGQDESSDKEITHSVMDSGRKEH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title