missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDB1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | DDB1 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18676215
|
Novus Biologicals
NBP2-98785-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633456
|
Novus Biologicals
NBP2-98785-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
DDB1 Polyclonal antibody specifically detects DDB1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifikationer
| DDB1 | |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Nucleotide Excision Repair | |
| 0.2 um filtered solution in PBS | |
| 1642 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| damage-specific DNA binding protein 1 (127kD), damage-specific DNA binding protein 1, 127kDa, Damage-specific DNA-binding protein 1, DDB p127 subunit, DDBa, DNA damage-binding protein 1, DNA damage-binding protein a, HBV X-associated protein 1, UV-damaged DNA-binding factor, UV-damaged DNA-binding protein 1, UV-DDB 1, UV-DDB1, XAP1, XAP-1, Xeroderma pigmentosum group E-complementing protein, XPCe, XPE, XPE-BF, XPE-binding factor | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human DDB1. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel