missing translation for 'onlineSavingsMsg'
Learn More

DDAH2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18387494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387494 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387494 Supplier Novus Biologicals Supplier No. NBP317699100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DDAH2 Polyclonal antibody specifically detects DDAH2 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH2
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias DDAHII, dimethylargininase-2, dimethylarginine dimethylaminohydrolase 2G6a, dimethylarginine dimethylaminohydrolase II, EC 3.5.3.18, G6a, N(G), N(G)-dimethylarginine dimethylaminohydrolase 2, NG30, NG-dimethylarginine dimethylamino hydrolase homolog, S-phase protein
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: ESLASGEGAGAGLPALDLAKAQREHGVLGGKLRQRLGLQLLELPPEESLPLGPLLGDTAVIQGDTALITRPWSPARRPEVDGVRKALQDLGLRIVEI
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23564
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.