missing translation for 'onlineSavingsMsg'
Learn More

DDAH2 Rabbit anti-Human, Mouse, Rat, Clone: 5M4Y4, Novus Biologicals™

Product Code. 18322133
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322133 100 μg 100µL
18365535 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18322133 Supplier Bio-Techne Supplier No. NBP316345100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

DDAH2 Monoclonal antibody specifically detects DDAH2 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DDAH2
Applications Western Blot
Classification Monoclonal
Clone 5M4Y4
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias DDAHII, dimethylargininase-2, dimethylarginine dimethylaminohydrolase 2G6a, dimethylarginine dimethylaminohydrolase II, EC 3.5.3.18, G6a, N(G), N(G)-dimethylarginine dimethylaminohydrolase 2, NG30, NG-dimethylarginine dimethylamino hydrolase homolog, S-phase protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human DDAH2 (O95865).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23564
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.