missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDAH2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38271-100ul
This item is not returnable.
View return policy
Description
DDAH2 Polyclonal antibody specifically detects DDAH2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| DDAH2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:100 | |
| DDAHII, dimethylargininase-2, dimethylarginine dimethylaminohydrolase 2G6a, dimethylarginine dimethylaminohydrolase II, EC 3.5.3.18, G6a, N(G), N(G)-dimethylarginine dimethylaminohydrolase 2, NG30, NG-dimethylarginine dimethylamino hydrolase homolog, S-phase protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-285 of human DDAH2 (NP_039268.1).,, Sequence:, MGTPGEGLGRCSHALIRGVPESLASGEGAGAGLPALDLAKAQREHGVLGGKLRQRLGLQLLELPPEESLPLGPLLGDTAVIQGDTALITRPWSPARRPEVDGVRKALQDLGLRIVEIGDENATLDGTDVLFTGREFFVGLSKWTNHRGAEIVADTFRDFAVSTVPVSGPSHLRGLCGMGGPRTVVAGSSDAAQKAVRAMAVLTDHPYASLTLPDDAAADCLFLRPGLPGVPPFLLHRGGGDLPNSQEALQKLSDVTLVPVSCSELEKAGAGLSSLCLVLSTRPHS | |
| 100 μL | |
| Signal Transduction | |
| 23564 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction