missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDA1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DDA1 |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16863954
|
Proteintech
14995-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16853954
|
Proteintech
14995-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May be involved in ubiquitination and subsequent proteasomal degradation of target proteins. Component of the DDD- E2 complexes which may provide a platform for interaction with CUL4A and WD repeat proteins.Specifications
| DDA1 | |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q9BW61, Q9D9Z5 | |
| 66498, 688813, 79016 | |
| DDA1 Fusion Protein Ag6981 | |
| Primary | |
| -20°C | |
| DDA1 |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C19orf58, DDA1, DET1 and DDB1 associated 1, PCIA 1, PCIA1 | |
| Dda1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title