missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCXR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15188-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene acts as a homotetramer to catalyze diacetyl reductase and L-xylulose reductase reactions. The encoded protein may play a role in the uronate cycle of glucose metabolism and in the cellular osmoregulation in the proximal renal tubules. Defects in this gene are a cause of pentosuria. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| DCXR | |
| Polyclonal | |
| Unconjugated | |
| DCXR | |
| Carbonyl reductase II, DCR, DCXR, HCR2, HCRII, KIDCR, Kidney dicarbonyl reductase, L xylulose reductase, P34H, SDR20C1, Sperm surface protein P34H, XR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 171408, 51181, 67880 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z4W1, Q91X52, Q920P0 | |
| Dcxr | |
| DCXR Fusion Protein Ag7272 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction