missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCXR Antibody (OTI7D11), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71906PCP
This item is not returnable.
View return policy
Description
DCXR Monoclonal antibody specifically detects DCXR in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| DCXR | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI7D11 | |
| Western Blot, Immunohistochemistry | |
| Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR | |
| Full length human recombinant protein of human DCXR (NP_057370) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Lipid and Metabolism | |
| 51181 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction