missing translation for 'onlineSavingsMsg'
Learn More

DCXR Antibody (6A6), Novus Biologicals™

Product Code. 18373699 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18373699 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18373699 Supplier Novus Biologicals Supplier No. H00051181M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DCXR Monoclonal antibody specifically detects DCXR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen DCXR
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6A6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057370
Gene Alias Carbonyl reductase II, DCR, dicarbonyl/L-xylulose reductaseP34H, EC 1.1.1, EC 1.1.1.10, HCR2, HCRII, human carbonyl reductase 2, KIDCR, Kidney dicarbonyl reductase, L-xylulose reductase, SDR20C1, short chain dehydrogenase/reductase family 20C, member 1, Sperm surface protein P34H, XR
Host Species Mouse
Immunogen DCXR (NP_057370, 145 a.a. ~ 244 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. NHSVYCSTKGALDMLTKVMALELGPHKIRVNAVNPTVVMTSMGQATWSDPHKAKTMLNRIPLGKFAEVEHVVNAILFLLSDRSGMTTGSTLPVEGGFWAC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Cytokine Research, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 51181
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.