Learn More
DCX Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13925-1-AP-20UL
 View more versions of this product
Product Code. 16872354
  • £135.00 / 20µL
Description
Anti-DCX antibodies are widely used for detection of neurogenesis. It is also a common neuron marker since its expression is specific to neurons.
In the developing cortex, cortical neurons must migrate over long distances to reach the site of their final differentiation. DCX is a cytoplasmic protein which appears to direct neuronal migration by regulating the organization and stability of microtubules. It contains two doublecortin domains, which bind microtubules. In addition, this protein interacts with LIS1, the regulatory gamma subunit of platelet activating factor acetylhydrolase, and this interaction is important to proper microtubule function in the developing cortex. Mutations in the gene encoding DCX are a cause of X-linked lissencephaly.
Specifications
Specifications
DCX
Polyclonal
Unconjugated
DCX
DBCN, DC, DCX, doublecortin, Doublin, Lis X, Lissencephalin X, LISX, SCLH, XLIS
Rabbit
Antigen Affinity Chromatography
RUO
13193, 1641
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43602, O88809
DCX
DCX Fusion Protein Ag4945
20 μL
Primary
Mouse, Human