missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCUN1D5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14810-1-AP-20UL
This item is not returnable.
View return policy
Description
Conjugation of Nedd8 to a cullin protein, termed neddylation, is an evolutionarily conserved process that functions to activate the cullin-RING family E3 ubiquitin ligases, leading to increased proteasomal degradation of a wide range of substrate proteins. Cellular neddylation requires the action of DCN1 (Defective in cullin neddylation protein 1), which, in humans, consists of five homologues designated as DCNL1-5. DCUN1D5 gene encodes the 28 kDa DCUN1 domain-containing protein 5 containing a C-terminal potentiating neddylation domain. The cellular function of DCUN1D5 needs to be explored, recently, a mono-ubiquitination-mediated mechanism that governs nuclear-cytoplasmic trafficking of hDCNL1, was reported to regulate hDCNL1-dependent activation of the cullin-RING E3 ubiquitin ligases in selected cellular compartments.Specifications
| DCUN1D5 | |
| Polyclonal | |
| Unconjugated | |
| DCUN1D5 | |
| DCN1 like protein 5, DCUN1D5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 315405, 76863, 84259 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot, Immunoprecipitation | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5PPL2, Q9BTE7, Q9CXV9 | |
| Dcun1d5 | |
| DCUN1D5 Fusion Protein Ag6534 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction