missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCUN1D3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | DCUN1D3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
DCUN1D3 Polyclonal specifically detects DCUN1D3 in Human samples. It is validated for Western Blot.Specifications
| DCUN1D3 | |
| Polyclonal | |
| Rabbit | |
| Q8IWE4 | |
| 123879 | |
| IgG | |
| 34 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| DCN1, defective in cullin neddylation 1, domain containing 3 (S. cerevisiae), DCN1-like protein 3, DCUN1 domain-containing protein 3, Defective in cullin neddylation protein 1-like protein 3, DKFZp686O0290, FLJ41725,44M2.4, MGC48972 | |
| DCUN1D3 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title