missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCUN1D2 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-97034CL1
This item is not returnable.
View return policy
Description
DCUN1D2 Polyclonal antibody specifically detects DCUN1D2 in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| DCUN1D2 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55208 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| C13orf17, chromosome 13 open reading frame 17, DCN1, defective in cullin neddylation 1, domain containing 2 (S. cerevisiae), DCN1-like protein 2, DCUN1 domain-containing protein 2, DCUN1L2, Defective in cullin neddylation protein 1-like protein 2, FLJ10704, FLJ20092 | |
| Produced in rabbits immunized with purified, recombinant Human DCUN1D2 (Accession#: NP_001014305.1; Met1-Phe259) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction