Learn More
DCUN1D2 Antibody (001), mFluor Violet 500 SE, Novus Biologicals™
Product Code. 18430429 Shop All Bio Techne Products
Product Code. 18430429 Supplier Novus Biologicals Supplier No. NBP290306MFV500
Rabbit Monoclonal Antibody
DCUN1D2 Monoclonal antibody specifically detects DCUN1D2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation