missing translation for 'onlineSavingsMsg'
Learn More

DcTRAILR2/TNFRSF22 Antibody (263310) [DyLight 755], Novus Biologicals™

Product Code. 30737025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737025 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737025 Supplier Novus Biologicals Supplier No. FAB1816Z

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

DcTRAILR2/TNFRSF22 Monoclonal antibody specifically detects DcTRAILR2/TNFRSF22 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen DcTRAILR2/TNFRSF22
Applications Western Blot
Classification Monoclonal
Clone 263310
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias APO-3;DDR3;DR3;GEF720;LARD;PLEKHG5;TNFRSF12;TNFRSF25;TR3;TRAMP;Tumor necrosis factor receptor superfamily member 25;WSL-1;WSL-LR
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R2/TNFRSF22, Ala39-Arg175, Accession # Q9ER62
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.