missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DcTRAILR1 Monoclonal Antibody (270028)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561149

Product Code. 30533811

  • £315.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

Mouse DcTRAILR1 and DcTRAILR2 (decoy TRAIL receptors 1 and 2) proteins are receptors for TRAIL according to ligand-receptor binding assays (Schneider et al. 2001). However, overexpression does not cause NF-kB activation or apoptosis, as would be expected of a TRAIL receptor. These observations led researchers to refer to these receptors as decoys. It is thought that cells that express decoy TRAIL receptor decoys at high levels relative to actual TRAIL receptors may use decoys as protection against the cytotoxic action of TRAIL (reviewed in Bouralexis et al. 2005). Additional analysis of these two murine TRAIL decoy receptors should help faciliate the understanding of the complex biology underlying TRAIL ligand/receptor interactions. DcTRAILR1 is an amino acid protein of 176 amino acids (GenBank no: NP-077252.2). DcTRAILR2 is an amino acid protein of 198 amino acids (GenBank no: AAN87807.1).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DcTRAILR1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9ER63
Tnfrsf23
Mouse myeloma cell line NS0-derived recombinant mouse DcTRAIL R1/TNFRSF23, Met30-Ser156.
100 μg
Primary
Mouse
Antibody
IgG2a
Western Blot
270028
Unconjugated
Tnfrsf23
Dctrailr1; Decoy TRAIL receptor 1; mDcTRAILR1; MGC123882; MGC123883; mSOB; SOB; TNF receptor family member SOB; TNF receptor homolog 1; Tnfrh1; Tnfrsf1al1; Tnfrsf23; TNFRSFH23; Tumor necrosis factor receptor p60 homolog 1; Tumor necrosis factor receptor superfamily member 23; tumor necrosis factor receptor superfamily, member 23
Rat
Protein A/G
RUO
79201
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.