missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCTN5 Recombinant Rabbit Monoclonal Antibody (24GB3540)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557022
This item is not returnable.
View return policy
Description
DCTN5 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a subunit of dynactin, a component of the cytoplasmic dynein motor machinery involved in minus-end-directed transport. The encoded protein is a component of the pointed-end subcomplex and is thought to bind membranous cargo. A pseudogene of this gene is located on the long arm of chromosome 1. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
Specifications
| DCTN5 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DCTN5 | |
| 4930427E12Rik; b2b315Clo; C78178; dctn5; dynactin 4; dynactin 5; dynactin 5 (p25); dynactin subunit 5; dynactin subunit 5-like protein; dynactin subunit p25; MGC3248 protein; p25; p25 dynactin subunit; RGD1305654; zgc:92842 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 308961, 59288, 84516 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3540 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BTE1, Q9QZB9 | |
| DCTN5 | |
| A synthesized peptide derived from human DCTN5 (aa 1-182). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction