missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCTN5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10182-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a subunit of dynactin, a component of the cytoplasmic dynein motor machinery involved in minus-end-directed transport. The encoded protein is a component of the pointed-end subcomplex and is thought to bind membranous cargo. A pseudogene of this gene is located on the long arm of chromosome 1. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Spécification
| DCTN5 | |
| Polyclonal | |
| Unconjugated | |
| DCTN5 | |
| DCTN5, dynactin 5 (p25), Dynactin subunit 5, Dynactin subunit p25, p25 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 308961, 59288, 84516 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BTE1, Q9QZB9 | |
| Dctn5 | |
| DCTN5 Fusion Protein Ag0234 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu