missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCTN4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | DCTN4 |
|---|---|
| Concentration | 0.19 mg/mL |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16836944
|
Proteintech
17098-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16826944
|
Proteintech
17098-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.Specifications
| DCTN4 | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| Q8CBY8, Q9QUR2, Q9UJW0 | |
| 51164, 67665, 84428 | |
| DCTN4 Fusion Protein Ag10579 | |
| Primary | |
| -20°C | |
| DCTN4 |
| 0.19 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DCTN4, dynactin 4 (p62), Dynactin subunit 4, Dynactin subunit p62 | |
| DCTN4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title