missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DCT Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552736

Product Code. 15700691

  • £438.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: WSGPYILRNQ DDRELWPRKF FHRTCKCTGN FAGYNCGDCK FGWTGPNCER KKPPVIRQNI HSLSPQEREQ FLGALDLAKK RVHPDYVITT QHWLGLLGPN GTQPQFANCS VYDF Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 46%.

The TYR-related proteins, TRP1 and TRP2, are also specifically expressed in melanocytes, and they likewise contribute to the synthesis of melanin within the melanosomes. The TRPs, including TYR, all share a similar transmembrane region, contain two metal-binding regions and a cysteinerich epidermal growth factor motif, and are localized in the melanosomal membrane. These proteins, however, have distinct catalytic activity, and they individually contribute to the biosynthesis of melanin biopolymers. The TRPs are believed to exist as a multi-enzyme complex, as these proteins form aggregates together, and the expression of TRP1 also helps stabilize TYR in melanocytes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DCT
Polyclonal
Unconjugated
DCT
Dct; DOPAchrome conversion factor; dopachrome delta isomerase; dopachrome delta-isomerase; DOPAchrome isomerase; DOPAchrome oxidoreductase; dopachrome tautomerase; dopachrome tautomerase (dopachrome delta-isomerase, tyrosine-related protein 2); DT; hypothetical protein LOC664745; L-dopachrome Delta-isomerase; L-dopachrome isomerase; L-dopachrome tautomerase; melanogenic enzyme; tyrosinase-related protein 2; trp2; RGD1564975; slaty; SLATY locus protein; slt; trp2; TRP-2; tyrosinase related protein 2; tyrosinase related protein-2; tyrosinase-related protein 2; tyrosinase-related protein-2; tyrosine-related protein 2; Tyrp2; Tyrp-2; zgc:109860
Rabbit
Antigen affinity chromatography
RUO
1638
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P40126
DCT
Recombinant protein corresponding to Human DCT. Recombinant protein control fragment (Product #RP-89997).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.