missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DCST2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | DCST2 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
DCST2 Polyclonal specifically detects DCST2 in Human samples. It is validated for Western Blot.Specifications
| DCST2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DC STAMP domain containing 2, DC-STAMP domain containing 2, DC-STAMP Domain-Containing Protein 2, FLJ32934 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of human DCST2 (NP_653223.2). Peptide sequence AQRKDPEQAWLLQQQLQEVLGRSLSMESTSESSDLDEEKGPQQRKHGQQP | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 127579 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title