missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DCST1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569232
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 79%; Dog: 79%; Guinea Pig: 93%; Horse: 79%; Human: 100%; Rabbit: 93%; Rat: 85%.
This gene encodes a protein with a domain similar to one found in dendritic cells (PMID:11169400) which play a key role in antigen processing and display for immune responses. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Sep 2011].
Specifications
| DCST1 | |
| Polyclonal | |
| Unconjugated | |
| DCST1 | |
| A330106H01Rik; Dcst1; DC-STAMP domain containing 1; DC-STAMP domain-containing protein 1; E3 ubiquitin-protein ligase DCST1; MLL5; RGD1307756; RING-type E3 ubiquitin transferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 149095 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q5T197 | |
| DCST1 | |
| synthetic peptide directed towards the C terminal region of human DCST1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction