missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DcR3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA587471

Product Code. 16555951

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: PLSTRVPGAE ECERAVIDFV AFQDISIKRL QRLLQALEAP EGWGPTPRAG RAALQLKLRR RLTELLGAQD GALLVRLLQA LRVARMPGLE RSVRERFLPV H Positive Samples: HT-29.

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DcR3
Polyclonal
Unconjugated
TNFRSF6B
DcR3; Decoy receptor 3; decoy receptor 3 variant 1; decoy receptor 3 variant 2; decoy receptor for Fas ligand; DJ583P15.1.1; hCG_2044836; M68; M68E; OTTHUMP00000031583; TNF receptor superfamily member 6b; TNFRSF6B; TR6; tumor necrosis factor receptor superfamily member 6B; tumor necrosis factor receptor superfamily member 6b, decoy; tumor necrosis factor receptor superfamily, member 6b, decoy; UNQ186/PRO212
Rabbit
Affinity Chromatography
RUO
8771
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1.19 mg/mL
PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
O95407
TNFRSF6B
Recombinant fusion protein containing a sequence corresponding to amino acids 200-300 of human TNFRSF6B (NP_0038141).
100 μL
Primary
Human, Rat, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.